Invitrogen
Manufacturer Code:PA576717
Catalog # PIPA576717
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human GHRH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GHRFGHRHGRFGrowth hormone releasing factorGrowth Hormone Releasing HormoneGrowth hormone-releasing factorGrowth hormone-releasing hormoneINNMGC119781OTTHUMP00000030915SermorelinSLIB&lowbarHUMANSomatocrininSomatoliberinSomatorelin; GHRH; GRF; Growth hormone-releasing factor; Growth hormone-releasing hormone; Sermorelin; Somatocrinin; Somatoliberin; Somatorelin
Gene Aliases: GHRF; GHRH; GRF; INN
UniProt ID: (Human) P01286, (Mouse) P16043
Entrez Gene ID: (Human) 2691, (Mouse) 14601
Molecular Function: neuropeptide peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.