Abnova
Manufacturer Code:PAB4992
Catalog # 89-131-674
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to human GHRH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of GHRH.
For Research Use Only
Protein Aliases: GHRF; GHRH; GRF; Growth hormone-releasing factor; Growth hormone-releasing hormone; MGC119781; Sermorelin; Somatocrinin; Somatoliberin; Somatorelin
Gene Aliases: GHRF; GHRH; GRF; INN
UniProt ID: (Human) P01286
Entrez Gene ID: (Human) 2691
Molecular Function: neuropeptide peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.