Invitrogen
Manufacturer Code:PA553035
Catalog # PIPA553035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human GGT7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gamma-glutamyltransferase 4; Gamma-glutamyltransferase 7; Gamma-glutamyltransferase 7 D20S101 GGTL5 dJ18C9.2 GGT4 GGTL3; Gamma-glutamyltransferase-like 3; Gamma-glutamyltransferase-like 5; Gamma-glutamyltranspeptidase 7; gamma-glutamyltranspeptidase-like 3; GGT 7; Glutathione hydrolase 7; Glutathione hydrolase 7 heavy chain; Glutathione hydrolase 7 light chain; mFLJ00311 protein
Gene Aliases: 1110017C11Rik; 6330563L03Rik; D20S101; dJ18C9.2; GGT4; GGT7; GGTL3; GGTL5
UniProt ID: (Human) Q9UJ14, (Mouse) Q99JP7, (Rat) Q99MZ4
Entrez Gene ID: (Human) 2686, (Mouse) 207182, (Rat) 156275
Molecular Function:
acyltransferase
hydrolase
protease
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.