Novus Biologicals
Manufacturer Code:NBP181883
Catalog # NBP181883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:DLAGDALLSLLAGDLGVEVPSAVPRPTLEPAEQLPVPQGILFTTPSPSAGPELLALLEAALRSGAPIPDPCPPFLQTAVSPESSALAAVDSSGSVLLLTSSLNCSFGSAHL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.3.2.2 FLJ25990 FLJ90165 gamma-glutamyltransferase 6 gamma-glutamyltransferase 6 homolog gamma-glutamyltransferase 6 homolog (rat) Gamma-glutamyltranspeptidase 6 GGT 6; Gamma-glutamyltransferase 6; gamma-glutamyltransferase 6 homolog; Gamma-glutamyltranspeptidase 6; GGT 6; Glutathione hydrolase 6; Glutathione hydrolase 6 heavy chain; Glutathione hydrolase 6 light chain
Gene Aliases: GGT6
UniProt ID: (Human) Q6P531
Entrez Gene ID: (Human) 124975
Molecular Function:
acyltransferase
hydrolase
protease
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.