Invitrogen
Manufacturer Code:PA572515
Catalog # PIPA572515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 130-156 amino acids from the N-terminal region of human GGTLA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gamma-glutamyl cleaving enzyme; Gamma-glutamyl leukotrienase; Gamma-glutamyl transpeptidase-related enzyme; gamma-glutamyl transpeptidase-related protein; Gamma-glutamyltransferase 5; Gamma-glutamyltransferase 5 GGT 5 Gamma-glutamyl transpeptidase-related enzyme GGT-rel Gamma-glutamyltransferase-like activity 1 Gamma-glutamyltranspeptidase 5 Glutathione hydrolase 5 Leukotriene-C4 hydrolase Gamma-glutamyltransferase 5 heavy chain Gamma-glutamyltransferase 5 light chain GGT5 GGTLA1; Gamma-glutamyltransferase-like activity 1; Gamma-glutamyltranspeptidase 5; GGL; GGT 5; GGT-rel; glutathione hydrolase 5; Glutathione hydrolase 5 heavy chain; Glutathione hydrolase 5 light chain; Glutathione hydrolase 5 proenzyme; Leukotriene-C4 hydrolase
Gene Aliases: AI551243; GGL; GGT 5; GGT-REL; GGT5; GGTLA1
UniProt ID: (Human) P36269, (Mouse) Q9Z2A9
Entrez Gene ID: (Human) 2687, (Mouse) 23887
Molecular Function:
acyltransferase
hydrolase
protease
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.