Novus Biologicals
Manufacturer Code:NBP297128G
Catalog # NB100986
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant GGT5 (NP_004112.2 Ser30-Tyr586). |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp566O011 EC 2.3.2.2 gamma-glutamyl cleaving enzyme Gamma-glutamyl transpeptidase-related enzyme gamma-glutamyl transpeptidase-related protein gamma-glutamyltransferase 5 Gamma-glutamyltransferase-like activity 1FLJ92733 Gamma-glutamyltranspeptidase 5 GGT 5 GGTLA1 GGT-rel; gamma-glutamyl cleaving enzyme; Gamma-glutamyl transpeptidase-related enzyme; gamma-glutamyl transpeptidase-related protein; Gamma-glutamyltransferase 5; Gamma-glutamyltransferase-like activity 1; Gamma-glutamyltranspeptidase 5; GGT 5; GGT-rel; glutathione hydrolase 5; Glutathione hydrolase 5 heavy chain; Glutathione hydrolase 5 light chain; Glutathione hydrolase 5 proenzyme; Leukotriene-C4 hydrolase
Gene Aliases: GGL; GGT 5; GGT-REL; GGT5; GGTLA1
UniProt ID: (Human) P36269
Entrez Gene ID: (Human) 2687
Molecular Function: acyltransferase hydrolase protease transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.