Invitrogen
Manufacturer Code:MA511815
Catalog # PIMA511815
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 138H11 |
Immunogen | Purified human kidney GGT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD224; CD224 D22S672 D22S732 GGT GGT 1 GTG gamma-glutamyl transpeptidase gamma-glutamyltranspeptidase 1 glutamyl transpeptidase; Gamma-glutamyltransferase 1; Gamma-glutamyltranspeptidase 1; GGT 1; glutathione hydrolase 1; Glutathione hydrolase 1 heavy chain; Glutathione hydrolase 1 light chain; Glutathione hydrolase 1 proenzyme; Leukotriene-C4 hydrolase; testicular tissue protein Li 73
Gene Aliases: CD224; D22S672; D22S732; GGT; GGT 1; GGT1; GTG
UniProt ID: (Human) P19440
Entrez Gene ID: (Human) 2678
Molecular Function:
acyltransferase
hydrolase
protease
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.