Novus Biologicals
Manufacturer Code:NBP298438R
Catalog # NB053853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant GGT1 (P07314 Pro27-Tyr568). |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD224; CD224 CD224 antigen D22S672 D22S732 EC 2.3.2.2 gamma-glutamyl transpeptidase gamma-glutamyltransferase 1MGC96904 gamma-glutamyltranspeptidase 1 GGT 1 GGTMGC96963 glutamyl transpeptidase GTG MGC96892; Gamma-glutamyltransferase 1; Gamma-glutamyltranspeptidase 1; GGT 1; glutathione hydrolase 1; Glutathione hydrolase 1 heavy chain; Glutathione hydrolase 1 light chain; Glutathione hydrolase 1 proenzyme; Leukotriene-C4 hydrolase; testicular tissue protein Li 73
Gene Aliases: CD224; D22S672; D22S732; GGT; GGT 1; GGT1; GTG
UniProt ID: (Human) P19440
Entrez Gene ID: (Human) 2678
Molecular Function:
acyltransferase
hydrolase
protease
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.