Abnova
Manufacturer Code:PAB27677
Catalog # 89-129-191
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of human GGT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of GGT1.
For Research Use Only
Protein Aliases: CD224; D22S672; D22S732; Gamma-glutamyltransferase 1; Gamma-glutamyltranspeptidase 1; GGT; GGT 1; glutathione hydrolase 1; Glutathione hydrolase 1 heavy chain; Glutathione hydrolase 1 light chain; Glutathione hydrolase 1 proenzyme; GTG; Leukotriene-C4 hydrolase; MGC96892; MGC96904; MGC96963; testicular tissue protein Li 73
Gene Aliases: CD224; D22S672; D22S732; GGT; GGT 1; GGT1; GTG
UniProt ID: (Human) P19440
Entrez Gene ID: (Human) 2678
Molecular Function:
acyltransferase
hydrolase
protease
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.