Invitrogen
Manufacturer Code:PA512168
Catalog # PIPA512168
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 270-300 amino acids from the C-terminal region of human GGPS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (2E,6E)-farnesyl diphosphate synthase; Dimethylallyltranstransferase; Farnesyl diphosphate synthase; Farnesyltranstransferase; Geranylgeranyl diphosphate synthase; Geranylgeranyl pyrophosphate synthase; geranylgeranyl pyrophosphate synthetase; Geranylgeranyl pyrophosphate synthetase GGPP synthetase GGPPSASE Geranylgeranyl diphosphate synthase; Geranyltranstransferase; GGPP synthase; GGPP synthetase; GGPPSase
Gene Aliases: 1810026C22Rik; 9530089B04Rik; AI843169; C79210; GGPPS; GGPPS1; GGPS1
UniProt ID: (Human) O95749, (Mouse) Q9WTN0
Entrez Gene ID: (Human) 9453, (Mouse) 14593
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.