Proteintech
Manufacturer Code:149441AP150UL
Catalog # 50-172-9853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GGPS1 Fusion Protein Ag6745 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (2E,6E)-farnesyl diphosphate synthase; Dimethylallyltranstransferase; Farnesyl diphosphate synthase; Farnesyltranstransferase; Geranylgeranyl diphosphate synthase; Geranylgeranyl pyrophosphate synthase; geranylgeranyl pyrophosphate synthetase; Geranyltranstransferase; GGPP synthase; GGPP synthase GGPPS GGPPS1 GGPPSase GGPS1; GGPP synthetase; GGPPSase
Gene Aliases: 1810026C22Rik; 9530089B04Rik; AI843169; C79210; Crlf3; GGPPS; GGPPS1; GGPS1; rGGPS1a; rGGPS1a1; rGGPS1a2; rGGPS1a3
UniProt ID: (Human) O95749, (Rat) Q6F596, (Mouse) Q9WTN0
Entrez Gene ID: (Human) 9453, (Rat) 291211, (Mouse) 14593
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.