Proteintech
Manufacturer Code:162091AP150UL
Catalog # 50-173-0792
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GGCX Fusion Protein Ag9185 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gamma glutamyl carboxylase GC GGCX VKCFD1; Gamma-glutamyl carboxylase; Peptidyl-glutamate 4-carboxylase; Vitamin K gamma glutamyl carboxylase; Vitamin K-dependent gamma-carboxylase
Gene Aliases: GC; GGCX; VKCFD1
UniProt ID: (Human) P38435, (Rat) O88496, (Mouse) Q9QYC7
Entrez Gene ID: (Human) 2677, (Rat) 81716, (Mouse) 56316
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.