Proteintech
Manufacturer Code:162571AP150UL
Catalog # 50-173-0833
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GGCT Fusion Protein Ag9284 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C7orf24 CRF21 Gamma glutamylcyclotransferase GCTG Ggc GGCT; Cytochrome c-releasing factor 21; gamma -glutamyl cyclotransferase; gamma-glutamyl cyclotransferase-like 1; Gamma-glutamylcyclotransferase
Gene Aliases: A030007L17Rik; AA673177; C7orf24; CRF21; GCTG; GGC; GGCT; Ggctl1; RGD1304876; RGD1562590
UniProt ID: (Human) O75223, (Mouse) Q9D7X8
Entrez Gene ID: (Human) 79017, (Mouse) 110175, (Rat) 362368
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.