Invitrogen
Manufacturer Code:PA528462
Catalog # PIPA528462
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 652 and 723 of Human GGA3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor-binding protein GGA3; ADP-ribosylation factor-binding protein GGA3 Golgi-localized gamma ear-containing ARF-binding protein 3 golgi associated gamma adaptin ear containing ARF binding protein 3; Golgi-localized, gamma ear-containing, ARF-binding protein 3
Gene Aliases: GGA3; KIAA0154
UniProt ID: (Human) Q9NZ52
Entrez Gene ID: (Human) 23163
Molecular Function: membrane traffic protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.