Novus Biologicals
Manufacturer Code:NBP271725AF405
Catalog # N271725A405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D2 |
Immunogen | Full length recombinant protein of human GGA2 (NP_055859) produced in HEK293T cells |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ADP-ribosylation factor-binding protein GGA2; FLJ20966 Gamma-adaptin-related protein 2 golgi associated gamma adaptin ear containing ARF binding protein 2 golgi-associated gamma adaptin ear containing ARF binding protein 2 Golgi-localized gamma ear-containing ARF-binding protein 2 KIAA1080ADP-ribosylation factor-binding protein GGA2 VEAR VHS domain and ear domain of gamma-adaptin VHS domain and ear domain-containing protein; Gamma-adaptin-related protein 2; golgi-associated, gamma adaptin ear containing, ARF binding protein 2; Golgi-localized, gamma ear-containing, ARF-binding protein 2; Vear; VHS domain and ear domain of gamma-adaptin; VHS domain and ear domain-containing protein
Gene Aliases: GGA2; KIAA1080; VEAR
UniProt ID: (Human) Q9UJY4
Entrez Gene ID: (Human) 23062
Molecular Function: membrane traffic protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.