Invitrogen
Manufacturer Code:PA545523
Catalog # PIPA545523
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human GGA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor binding protein 2; ADP-ribosylation factor-binding protein GGA2; Gamma-adaptin-related protein 2; golgi-associated gamma adaptin ear containing ARF binding protein 2 VEAR; golgi-associated, gamma adaptin ear containing, ARF binding protein 2; Golgi-localized, gamma ear-containing, ARF-binding protein 2; Vear; VHS domain and ear domain of gamma-adaptin; VHS domain and ear domain-containing protein
Gene Aliases: 1200007E24Rik; GGA2; KIAA1080; mKIAA1080; VEAR
UniProt ID: (Human) Q9UJY4, (Mouse) Q6P5E6
Entrez Gene ID: (Human) 23062, (Dog) 608555, (Rabbit) 100355285, (Horse) 100069034, (Mouse) 74105, (Rat) 293455, (Guinea pig) 100720654
Molecular Function: membrane traffic protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.