Abnova
Manufacturer Code:H00002674M05
Catalog # 89-022-302
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4D8 |
Immunogen | GFRA1 (NP_005255 32 a.a. ~ 119 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant GFRA1.
For Research Use Only
Protein Aliases: GDNF family receptor alpha-1; GDNF receptor alpha-1; GDNFR; GDNFR-alpha-1; GDNFRA; GFR-ALPHA-1; Glial cell line-derived neurotrophic factor receptor alpha; GPI-linked anchor protein; MGC23045; PI-linked cell-surface accessory protein; RET ligand 1; RET1L; RETL1; TGF-beta-related neurotrophic factor receptor 1; TRNR1
Gene Aliases: GDNFR; GDNFRA; GFR-ALPHA-1; GFRA1; RET1L; RETL1; TRNR1
UniProt ID: (Human) P56159
Entrez Gene ID: (Human) 2674
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.