Invitrogen
Manufacturer Code:PA577540
Catalog # PIPA577540
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)GSRDG(S)PEEGGPR corresponding to amino acid residues 160-172 of rat GFRalpha4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDNF family receptor alpha-4; GDNF receptor alpha-4; GDNFR-alpha-4; GFR alpha-4; GFR-alpha-4; GFRA4; glial cell line derived neurotrophic factor family receptor alpha 4; neurotrophic factor receptor splice variant A (Gfra4 gene); Persephin receptor
Gene Aliases: AW060733; G630015H18Rik; Gfra4
UniProt ID: (Rat) Q9EPI2, (Mouse) Q9JJT2
Entrez Gene ID: (Rat) 66023, (Mouse) 14588
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.