Novus Biologicals
Manufacturer Code:NBP179553
Catalog # NBP179553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C terminal of human GFRA2. Peptide sequence NVSPKGPSFQATQAPRVEKTPSLPDDLSDSTSLGTSVITTCTSVQEQGLK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDNF family receptor alpha 2 GDNF family receptor alpha-2 GDNF receptor alpha-2 GDNF receptor beta GDNFR-alpha-2 GDNFR-beta GDNFRBRET ligand 2 GFR-alpha 2 GFR-alpha-2 glial cell line derived neurotrophic factor receptor beta Neurturin receptor alpha NRTNR-alpha NTNRA NTNR-alpha PI-linked cell-surface accessory protein RETL2TGF-beta-related neurotrophic factor receptor 2 TGF-beta related neurotrophic factor receptor 2 TRN receptor GPI-anchored TRNR2NRTNR-ALPHA; GDNF family receptor alpha-2; GDNF receptor alpha-2; GDNF receptor beta; GDNFR-alpha-2; GDNFR-beta; GFR-alpha 2; glial cell line derived neurotrophic factor receptor, beta; Neurturin receptor alpha; NRTNR-alpha; NTNR-alpha; PI-linked cell-surface accessory protein; RET ligand 2; TGF-beta-related neurotrophic factor receptor 2; TRN receptor, GPI-anchored
Gene Aliases: GDNFRB; GFRA2; NRTNR-ALPHA; NTNRA; RETL2; TRNR2
UniProt ID: (Human) O00451
Entrez Gene ID: (Human) 2675
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.