Invitrogen
Manufacturer Code:PA5115280
Catalog # PIPA5115280
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GDNF Receptor alpha 2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDNF family receptor alpha 2 GDNF family receptor alpha-2 GDNF re GDNF receptor alpha-2 GDNF receptor beta GDNFR-alpha-2 Gdnfrb GDNFR-beta GFRA2 GFR-alpha 2 GFR-alpha-2 glial cell line derived neurotrophic factor family receptor alpha 2 glial cell line derived neurotrophic factor receptor beta HpV18gp1 Neurturin receptor alpha NRTNR-ALPHA NTNRA NTNR-alpha PI-linked cell-surface accessory protein RET ligand 2 Retl2 TGF-beta-related neurotrophic factor receptor 2 TRN receptor GPI-anchored Trnr2 tyrosine kinase receptor ligand 2; GDNF family receptor alpha-2; GDNF receptor alpha-2; GDNF receptor beta; GDNFR-alpha-2; GDNFR-beta; GFR-alpha 2; glial cell line derived neurotrophic factor family receptor alpha 2; glial cell line derived neurotrophic factor receptor, beta; Neurturin receptor alpha; NRTNR-alpha; NTNR-alpha; PI-linked cell-surface accessory protein; RET ligand 2; TGF-beta-related neurotrophic factor receptor 2; TRN receptor, GPI-anchored; tyrosine kinase receptor ligand 2
Gene Aliases: GDNFRB; GFRA2; NRTNR-ALPHA; NTNRA; RETL2; TRNR2
UniProt ID: (Human) O00451, (Mouse) O08842
Entrez Gene ID: (Human) 2675, (Rat) 25136, (Mouse) 14586
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.