Invitrogen
Manufacturer Code:PA118399
Catalog # 01-670-650
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues P(376) R V E K T P S L P D D L S D(390) of human GFR alpha-2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDNF family receptor alpha-2; GDNF family receptor alpha-2 GFR-alpha-2 Neurturin receptor alpha NTNR-alpha NRTNR-alpha TGF-beta-related neurotrophic factor receptor 2 GDNF re; glial cell line derived neurotrophic factor family receptor alpha 2; tyrosine kinase receptor ligand 2
Gene Aliases: Retl2
Entrez Gene ID: (Rat) 25136
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.