R&D Systems
Manufacturer Code:MAB560SP
Catalog # MAB560SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 81401 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant rat GFR alpha-1/GDNF R alpha-1 Asp25-Leu445 Accession # Q62997 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp313E1012 DKFZp686J0156 GDNF family receptor alpha 1 GDNF family receptor alpha-1 GDNF R alpha-1 GDNF receptor alpha-1 GDNFRAGFR-ALPHA-1 GDNFR-alpha-1 GDNFRMGC23045 GFR alpha1 GFRa-1 GFRA1 GFR-alpha-1 Glial cell line-derived neurotrophic factor receptor alpha GPI-linked anchor protein PI-linked cell-surface accessory protein RET ligand 1 RET1L RETL1FLJ10561 TGF-beta related neurotrophic factor receptor 1 TGF-beta-related neurotrophic factor receptor 1 TRNR1FLJ31546; GDNF family receptor alpha-1; GDNF receptor alpha-1; GDNFR-alpha-1; Glial cell line-derived neurotrophic factor receptor alpha; GPI-linked anchor protein; PI-linked cell-surface accessory protein; RET ligand 1; TGF-beta-related neurotrophic factor receptor 1
Gene Aliases: GDNFR; GDNFRA; GFR-ALPHA-1; GFRA1; RET1L; RETL1; TRNR1
UniProt ID: (Human) P56159
Entrez Gene ID: (Human) 2674
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.