Invitrogen
Manufacturer Code:MA529712
Catalog # PIMA529712
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 9 |
Immunogen | Recombit Mouse GFRA1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp313E1012 DKFZp686J0156 FLJ10561 FLJ31546 GDNFR GDNFRA GFR-ALPHA-1 MGC23045 RET1L RETL1 TRNR1; GDNF family receptor alpha-1; GDNF receptor alpha-1; GDNFR-alpha; GDNFR-alpha-1; GFR alpha-1; GFR-alpha-1; TGF-beta-related neurotrophic factor receptor 1
Gene Aliases: AU042498; Gdnfra; Gfra1; Trnr1
UniProt ID: (Mouse) P97785
Entrez Gene ID: (Mouse) 14585
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.