Invitrogen
Manufacturer Code:MA530610
Catalog # PIMA530610
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 29 |
Immunogen | Recombit Rat GFRA1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp313E1012 DKFZp686J0156 FLJ10561 FLJ31546 GDNFR GDNFRA GFR-ALPHA-1 MGC23045 RET1L RETL1 TRNR1; GDNF family receptor alpha-1; GDNF receptor alpha; GDNF receptor alpha-1; GDNFR-alpha; GDNFR-alpha-1; GFR-alpha-1; glial cell line derived neurotrophic factor family receptor alpha 1; Glial cell line-derived neurotrophic factor receptor alpha; RET ligand 1; TGF-beta-related neurotrophic factor receptor 1
Gene Aliases: Gdnfra; Gfra1; Retl1; Trnr1
UniProt ID: (Rat) Q62997
Entrez Gene ID: (Rat) 25454
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.