Novus Biologicals
Manufacturer Code:NBP252474
Catalog # NBP252474
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F1B9 |
Immunogen | Purified recombinant fragment of human GFPT1 (AA: 536-681) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: D-fructose-6-phosphate amidotransferase 1; GFAT 1; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1 CMSTA1 GFA GFAT GFAT 1 GFAT1 GFAT1m GFPT GFPT1L glutamine--fructose-6-phosphate transaminase 1 MSLG; Glutamine--fructose-6-phosphate aminotransferase [isomerizing] 1; Glutamine:fructose-6-phosphate amidotransferase 1; Hexosephosphate aminotransferase 1
Gene Aliases: CMS12; CMSTA1; GFA; GFAT; GFAT 1; GFAT1; GFAT1m; GFPT; GFPT1; GFPT1L; MSLG
UniProt ID: (Human) Q06210
Entrez Gene ID: (Human) 2673
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.