Invitrogen
Manufacturer Code:PA532776
Catalog # PIPA532776
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 12 amino acid peptide from N-Terminus of human GFPT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D-fructose-6-phosphate amidotransferase 1; GFA GFAT GFAT 1 GFAT1 GFAT1m GFPT GFPT1L MSLG D-fructose-6-phosphate amidotransferase 1 glucosamine--fructose-6-phosphate aminotransferase &lsqbisomerizing] 1 glutamine:fructose 6 phosphate amidotransferase 1 hexosephosphate aminotransferase 1; GFAT 1; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1; Glutamine--fructose-6-phosphate aminotransferase [isomerizing] 1; Glutamine:fructose-6-phosphate amidotransferase 1; Hexosephosphate aminotransferase 1
Gene Aliases: CMS12; CMSTA1; GFA; GFAT; GFAT 1; GFAT1; GFAT1m; GFPT; GFPT1; GFPT1L; MSLG
UniProt ID: (Human) Q06210
Entrez Gene ID: (Human) 2673
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.