Invitrogen
Manufacturer Code:PA5103033
Catalog # PIPA5103033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GFPT1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810423A18Rik AI324119 AI449986 CMS12 CMSTA1 D-fructose-6-phosphate amidotransferase 1 eartha GFA GFAT GFAT 1 GFAT1 GFAT1m GFPT GFPT1 GFPT1L glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1 glutamine fructose-6-phosphate transaminase 1 glutamine:fructose 6 phosphate amidotransferase 1 Glutamine:fructose-6-phosphate amidotransferase 1 glutamine--fructose-6-phosphate aminotransferase [isomerizing] 1 glutamine--fructose-6-phosphate transaminase 1 harvest moon hexosephosphate aminotransferase 1 hmn MSLG unm_z110; D-fructose-6-phosphate amidotransferase 1; GFAT 1; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1; Glutamine--fructose-6-phosphate aminotransferase [isomerizing] 1; glutamine:fructose 6 phosphate amidotransferase 1; Glutamine:fructose-6-phosphate amidotransferase 1; Hexosephosphate aminotransferase 1
Gene Aliases: CMS12; CMSTA1; GFA; GFAT; GFAT 1; GFAT1; GFAT1m; GFPT; GFPT1; GFPT1L; MSLG
UniProt ID: (Human) Q06210, (Rat) P82808
Entrez Gene ID: (Human) 2673, (Rat) 297417
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.