Novus Biologicals
Manufacturer Code:NBP299692B
Catalog # NB088985
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant GFPT1 (AAA58502.1 Gln 332-Glu 699). |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D-fructose-6-phosphate amidotransferase 1; GFAT 1; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1 CMSTA1 GFA GFAT GFAT 1 GFAT1 GFAT1m GFPT GFPT1L glutamine--fructose-6-phosphate transaminase 1 MSLG; Glutamine--fructose-6-phosphate aminotransferase [isomerizing] 1; Glutamine:fructose-6-phosphate amidotransferase 1; Hexosephosphate aminotransferase 1
Gene Aliases: CMS12; CMSTA1; GFA; GFAT; GFAT 1; GFAT1; GFAT1m; GFPT; GFPT1; GFPT1L; MSLG
UniProt ID: (Human) Q06210
Entrez Gene ID: (Human) 2673
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.