Novus Biologicals
Manufacturer Code:NBP19845820UL
Catalog # NBP19845820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is Gfm1 - C-terminal region. Peptide sequence ELRSCTEGKGEYTMEYCRYQPCSPSTQEELINKYLEATGQLPVKKGKAKN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EF-Gmt; EFG EFG1FLJ12662 EFGM EF-Gmt EGF1 FLJ13632 FLJ20773 G elongation factor mitochondrial 1 G translation elongation factor mitochondrial GFMCOXPD1 hEFG1 mEF-G 1 mitochondrial mitochondrial elongation factor G mitochondrial elongation factor G1; Elongation factor G 1, mitochondrial; Elongation factor G, mitochondrial; Elongation factor G1; G elongation factor, mitochondrial 1; G translation elongation factor, mitochondrial; GFM1; hEFG1; mEF-G 1; mitochondrial elongation factor G; mitochondrial elongation factor G1
Gene Aliases: COXPD1; EFG; EFG1; EFGM; EGF1; GFM; GFM1; hEFG1
UniProt ID: (Human) Q96RP9
Entrez Gene ID: (Human) 85476
Molecular Function: G-protein RNA binding protein enzyme modulator hydrolase nucleic acid binding translation elongation factor translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.