Abnova
Manufacturer Code:H00085476K
Catalog # 89-203-698
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human GFM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human GFM1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: COXPD1 EFG EFG1 EFGM EGF1 FLJ12662 FLJ13632 FLJ20773 GFM hEFG1; EF-Gmt; Elongation factor G 1, mitochondrial; Elongation factor G, mitochondrial; Elongation factor G1; G elongation factor, mitochondrial 1; G translation elongation factor, mitochondrial; GFM1; hEFG1; mEF-G 1; mitochondrial elongation factor G; mitochondrial elongation factor G1
Gene Aliases: COXPD1; EFG; EFG1; EFGM; EGF1; GFM; GFM1; hEFG1
UniProt ID: (Human) Q96RP9
Entrez Gene ID: (Human) 85476
Molecular Function: G-protein RNA binding protein enzyme modulator hydrolase nucleic acid binding translation elongation factor translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.