OriGene
Manufacturer Code:CF805544
Catalog # 50-166-7472
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6E8 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-180 of human GFI1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: GFI-1 GFI1A SCN2 ZNF163; growth factor independence-1; growth factor independent 1 transcription repressor; Growth factor independent protein 1; Zinc finger protein 163; Zinc finger protein Gfi-1
Gene Aliases: GFI-1; GFI1; GFI1A; SCN2; ZNF163
UniProt ID: (Human) Q99684
Entrez Gene ID: (Human) 2672
Molecular Function:
DNA binding protein
centromere DNA-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.