Invitrogen
Manufacturer Code:PA523495
Catalog # PIPA523495
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Portion of amino acid 280-330 of human Zinc finger protein Gfi-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GFI-1 SCN2 ZNF163 growth factor independence-1 growth factor independent protein 1 zinc finger protein 163 zinc finger protein Gfi-1; growth factor independence-1; growth factor independent 1 transcription repressor; Growth factor independent protein 1; Zinc finger protein 163; Zinc finger protein Gfi-1
Gene Aliases: AW495828; GFI-1; GFI1; GFI1A; Pal-1; Pal1; SCN2; ZNF163
UniProt ID: (Human) Q99684, (Mouse) P70338
Entrez Gene ID: (Human) 2672, (Mouse) 14581
Molecular Function:
DNA binding protein
centromere DNA-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.