Invitrogen
Manufacturer Code:PA5116697
Catalog # PIPA5116697
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human GFI1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW495828 FLJ94509 GFI1 GFI-1 GFI1A growth factor independence-1 growth factor independent 1 growth factor independent 1 transcription repressor growth factor independent 1 transcriptional repressor growth factor independent protein 1 Growth factor independent-1 Pal1 Pal-1 SCN2 Zinc finger protein 163 zinc finger protein Gfi-1 ZNF163; growth factor independence-1; growth factor independent 1 transcription repressor; Growth factor independent protein 1; Growth factor independent-1; Zinc finger protein 163; Zinc finger protein Gfi-1
Gene Aliases: AW495828; GFI-1; GFI1; GFI1A; Pal-1; Pal1; SCN2; ZNF163
UniProt ID: (Human) Q99684, (Rat) Q07120, (Mouse) P70338
Entrez Gene ID: (Human) 2672, (Rat) 24388, (Mouse) 14581
Molecular Function: DNA binding protein centromere DNA-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.