Proteintech
Manufacturer Code:239351AP150UL
Catalog # 50-173-4168
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GFAP Fusion Protein Ag20853 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GFAP; Glial fibrillary acidic protein; glial fibrillary acidic protein alpha; intermediate filament; intermediate filament protein
Gene Aliases: AI836096; ALXDRD; GFAP
UniProt ID: (Human) Q9UFD0, (Rat) P47819, (Mouse) P03995
Entrez Gene ID: (Human) 2670, (Rat) 24387, (Mouse) 14580
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.