Novus Biologicals
Manufacturer Code:NBP186732
Catalog # NBP186732
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:AVDGGKLTVFTVLCEQYQPSLRRDPMYNEYLDRIGQLFFGVPPKQTSSYGGLLGNLLTSLMGSSEQEDGEESPSDGSPIELD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CEEchromosome 7 open reading frame 20 CGI-20 golgi to ER traffic protein 4 homolog (S. cerevisiae) TRC3535kDa; conserved edge expressed protein; conserved edge protein; Conserved edge-expressed protein; Golgi to ER traffic protein 4 homolog; H_NH1244M04.5; Transmembrane domain recognition complex 35 kDa subunit; transmembrane domain recognition complex, 35kDa; TRC35
Gene Aliases: C7orf20; CEE; CGI-20; GET4; TRC35
UniProt ID: (Human) Q7L5D6
Entrez Gene ID: (Human) 51608
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.