Invitrogen
Manufacturer Code:PA526502
Catalog # PIPA526502
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 60-89 amino acids from the N-terminal region of human GEN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Flap endonuclease GEN homolog 1; Gen endonuclease homolog 1; Gen Gen homolog 1 endonuclease flap endonuclease GEN homolog 1; Gen homolog 1, endonuclease; GEN1 Holliday junction 5' flap endonuclease; Holliday junction resolvase
Gene Aliases: 5830483C08Rik; Gen; GEN1
UniProt ID: (Human) Q17RS7, (Mouse) Q8BMI4
Entrez Gene ID: (Human) 348654, (Mouse) 209334
Molecular Function:
DNA binding protein
damaged DNA-binding protein
endodeoxyribonuclease
exodeoxyribonuclease
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.