Novus Biologicals
Manufacturer Code:NBP230783
Catalog # NBP230783
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: SILSVKESCIANSGSDCTSHLSKDLPGIPLQNESRDSKILKGDQLLQEDYKVNTSVPYSVSNTVVKTCNVRPPNTALDHSRKVDMQTTRKILMKKSVCLDRHSSDEQSAPVFGKAKYTTQRMKHSSQKHNSSHFKESGHNKLSSPKI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp781F0986 EC 3.1 flap endonuclease GEN homolog 1 FLJ40869 Gen Gen homolog 1 endonuclease (Drosophila); Flap endonuclease GEN homolog 1; Gen endonuclease homolog 1; Gen homolog 1, endonuclease; GEN1 Holliday junction 5' flap endonuclease; Holliday junction resolvase
Gene Aliases: Gen; GEN1
UniProt ID: (Human) Q17RS7
Entrez Gene ID: (Human) 348654
Molecular Function:
DNA binding protein
damaged DNA-binding protein
endodeoxyribonuclease
exodeoxyribonuclease
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.