Invitrogen
Manufacturer Code:MA534750
Catalog # PIMA534750
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG36-46 |
Immunogen | Recombinant protein corresponding to C-terminal human GEF H1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AA408978 Arhgef2 CG10188 DKFZp547L106 DKFZp547P1516 DmelCG10188 Dmel_CG10188 FLJ45440 GEF GEFH1 GEF-H1 Guanine nucleotide exchange factor H1 KIAA0651 Lbcl1 LBC&primeS first cousin Lfc LFP40 Lymphoid blast crisis-like 1 Microtubule-regulated Rho-GEF mKIAA0651 Oncogene LFC OTTHUMP00000018855 P40 Proliferating cell nucleolar antigen p40 Rho guanine nucleotide exchange factor 2 Rhobin; GEF-H1; Guanine nucleotide exchange factor H1; Microtubule-regulated Rho-GEF; Proliferating cell nucleolar antigen p40; Rho guanine nucleotide exchange factor 2; Rho/Rac guanine nucleotide exchange factor (GEF) 2
Gene Aliases: ARHGEF2; GEF; GEF-H1; GEFH1; KIAA0651; LFP40; P40
UniProt ID: (Human) Q8WUG4
Entrez Gene ID: (Human) 9181
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.