Invitrogen
Manufacturer Code:MA527803
Catalog # PIMA527803
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT336 |
Immunogen | Recombit protein encompassing a sequence within the C-terminus region of human GEF-H1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp547L106 DKFZp547P1516 FLJ45440 GEF GEF-H1 GEFH1 KIAA0651 LFP40 P40 ARHGEF2 microtubule-regulated Rho-GEF Rho/Rac guanine nucleotide exchange factor 2 proliferating cell nucleolar antigen p40 rho guanine nucleotide exchange factor 2 guanine nucleotide exchange factor H1 Rho/Rac guanine nucleotide exchange factor (GEF) 2; GEF-H1; Guanine nucleotide exchange factor H1; LBC'S first cousin; Lymphoid blast crisis-like 1; Microtubule-regulated Rho-GEF; Oncogene LFC; Proliferating cell nucleolar antigen p40; Rho guanine nucleotide exchange factor 2; Rho/Rac guanine nucleotide exchange factor (GEF) 2; Rhobin
Gene Aliases: AA408978; ARHGEF2; GEF; GEF-H1; GEFH1; KIAA0651; Lbcl1; Lfc; LFP40; mKIAA0651; P40
UniProt ID: (Human) Q8WUG4, (Mouse) Q60875, (Rat) Q5FVC2
Entrez Gene ID: (Human) 9181, (Mouse) 16800, (Rat) 310635
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.