Novus Biologicals
Manufacturer Code:NBP243584AF532
Catalog # NP24384A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1132 |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human GEF-H1. The exact sequence is proprietary. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: GEF GEFH1 GEF-H1DKFZp547P1516 Guanine nucleotide exchange factor H1 KIAA0651DKFZp547L106 LFP40FLJ45440 Microtubule-regulated Rho-GEF P40 Proliferating cell nucleolar antigen p40 rho guanine nucleotide exchange factor 2 Rho/Rac guanine nucleotide exchange factor (GEF) 2 Rho/Rac guanine nucleotide exchange factor 2; GEF-H1; Guanine nucleotide exchange factor H1; Microtubule-regulated Rho-GEF; Proliferating cell nucleolar antigen p40; Rho guanine nucleotide exchange factor 2; Rho/Rac guanine nucleotide exchange factor (GEF) 2
Gene Aliases: ARHGEF2; GEF; GEF-H1; GEFH1; KIAA0651; LFP40; P40
UniProt ID: (Human) Q8WUG4
Entrez Gene ID: (Human) 9181
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.