Invitrogen
Manufacturer Code:PA564287
Catalog # PIPA564287
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human GDPD5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDE2 PP1665 GDE2 PP6037 PP9363 UNQ1850/PRO3580 Glycerophosphodiester phosphodiesterase domain-containing protein 5 Glycerophosphodiester phosphodiesterase 2; Glycerophosphocholine phosphodiesterase GDPD5; Glycerophosphodiester phosphodiesterase 2; Glycerophosphodiester phosphodiesterase domain-containing protein 5; Phosphoinositide phospholipase C GDPD5
Gene Aliases: GDE2; GDPD5; PP1665; PP6037; PP9363; UNQ1850/PRO3580
UniProt ID: (Human) Q8WTR4
Entrez Gene ID: (Human) 81544
Molecular Function:
esterase
hydrolase
ligase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.