Invitrogen
Manufacturer Code:PA543468
Catalog # PIPA543468
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human GDPD5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycerophosphocholine phosphodiesterase GDPD5; Glycerophosphodiester phosphodiesterase 2; glycerophosphodiester phosphodiesterase domain containing 5; Glycerophosphodiester phosphodiesterase domain-containing protein 5; Phosphoinositide phospholipase C GDPD5
Gene Aliases: BC024955; GDE2; GDPD5; PP1665; PP6037; PP9363; RGD1559673; UNQ1850/PRO3580
UniProt ID: (Human) Q8WTR4, (Mouse) Q640M6
Entrez Gene ID: (Dog) 485188, (Bovine) 532719, (Rabbit) 100355595, (Human) 81544, (Pig) 100627912, (Horse) 100064592, (Mouse) 233552, (Guinea pig) 100728386, (Rat) 499211
Molecular Function:
esterase
hydrolase
ligase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.