Invitrogen
Manufacturer Code:PA538048
Catalog # PIPA538048
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | 18 amino acid peptide near the amino terminus of human GDPD5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDE2 PP1665 GDE2 PP6037 PP9363 UNQ1850/PRO3580 Glycerophosphodiester phosphodiesterase domain-containing protein 5 Glycerophosphodiester phosphodiesterase 2; Glycerophosphocholine phosphodiesterase GDPD5; Glycerophosphodiester phosphodiesterase 2; Glycerophosphodiester phosphodiesterase domain-containing protein 5; Phosphoinositide phospholipase C GDPD5
Gene Aliases: BC024955; GDE2; GDPD5; PP1665; PP6037; PP9363; RGD1559673; UNQ1850/PRO3580
UniProt ID: (Human) Q8WTR4, (Mouse) Q640M6
Entrez Gene ID: (Human) 81544, (Mouse) 233552, (Rat) 499211
Molecular Function: esterase hydrolase ligase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.