Novus Biologicals
Manufacturer Code:NBP297884IR
Catalog # NB048936
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human GDPD3 fragment. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1 FLJ22603 glycerophosphodiester phosphodiesterase domain containing 3 glycerophosphodiester phosphodiesterase domain-containing protein 3 MGC4171; Glycerophosphodiester phosphodiesterase 7; Glycerophosphodiester phosphodiesterase domain-containing protein 3; Lysophospholipase D GDPD3
Gene Aliases: GDE7; GDPD3
UniProt ID: (Human) Q7L5L3
Entrez Gene ID: (Human) 79153
Molecular Function: esterase hydrolase ligase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.