Abnova
Manufacturer Code:PAB23807
Catalog # 89-125-515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human GDPD3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant GDPD3.
For Research Use Only
Protein Aliases: FLJ22603; Glycerophosphodiester phosphodiesterase 7; Glycerophosphodiester phosphodiesterase domain-containing protein 3; Lysophospholipase D GDPD3; MGC4171
Gene Aliases: GDE7; GDPD3
UniProt ID: (Human) Q7L5L3
Entrez Gene ID: (Human) 79153
Molecular Function: esterase hydrolase ligase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.