Invitrogen
Manufacturer Code:MA524213
Catalog # PIMA524213
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 501016 |
Immunogen | HEK293 human embryonic kidney cell line transfected with human GDPD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: GDE3 OBDPF EC 3.1.4.43 FLJ20207 GDE3Osteoblast differentiation promoting factor Glycerophosphodiester phosphodiesterase 3 glycerophosphodiester phosphodiesterase domain containing 2 Glycerophosphodiester phosphodiesterase domain-containing protein 2 OBDPFglycerophosphoinositol inositolphosphodiesterase GDPD2; Glycerophosphodiester phosphodiesterase 3; Glycerophosphodiester phosphodiesterase domain-containing protein 2; Glycerophosphoinositol inositolphosphodiesterase GDPD2; Osteoblast differentiation promoting factor
Gene Aliases: 9130017L10Rik; GDE3; GDPD2; OBDPF; UNQ1935/PRO4418
UniProt ID: (Human) Q9HCC8, (Mouse) Q9ESM6
Entrez Gene ID: (Human) 54857, (Mouse) 71584
Molecular Function: esterase hydrolase ligase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.