Novus Biologicals
Manufacturer Code:NBP212335AF532
Catalog # NP21233A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0308-12F9 |
Immunogen | Human GDNF |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Astrocyte-derived trophic factor; Astrocyte-derived trophic factor ATF ATF1 ATF2 glial cell derived neurotrophic factor glial cell line derived neurotrophic factor glial cell line-derived neurotrophic factor HFB1-GDNF hGDNF HSCR3; ATF; Glial cell line-derived neurotrophic factor; hGDNF
Gene Aliases: ATF1; ATF2; GDNF; HFB1-GDNF; HSCR3
UniProt ID: (Human) P39905
Entrez Gene ID: (Human) 2668
Molecular Function: growth factor neurotrophic factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.