Invitrogen
Manufacturer Code:711074
Catalog # PI711074
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 13HCLC |
Immunogen | Protein corresponding to Human GDNF (aa 78-211) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: Astrocyte-derived trophic factor; ATF; Glial Cell Derived Neurotrophic Factor; Glial cell line-derived neurotrophic factor; hGDNF
Gene Aliases: ATF1; ATF2; GDNF; HFB1-GDNF; HSCR3
UniProt ID: (Human) P39905
Entrez Gene ID: (Human) 2668
Molecular Function: growth factor neurotrophic factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.