Invitrogen
Manufacturer Code:PA594994
Catalog # PIPA594994
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human GDNF different from the related mouse sequence by three amino acids and rat sequence by two amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Astrocyte-derived trophic factor; ATF; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; Glial cell line-derived neurotrophic factor; Glial cell line-derived neurotrophic factor hGDNF Astrocyte-derived trophic factor ATF GDNF; hGDNF; mGDNF; neurotrophic factor
Gene Aliases: AI385739; ATF1; ATF2; GDNF; gndf; HFB1-GDNF; HSCR3
UniProt ID: (Human) P39905, (Mouse) P48540, (Rat) Q07731
Entrez Gene ID: (Human) 2668, (Mouse) 14573, (Rat) 25453
Molecular Function: growth factor neurotrophic factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.