R&D Systems
Manufacturer Code:AF212SP
Catalog # AF212SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived and Mouse myeloma cell line NS0-derived recombinant human GDNF Arg109-Ile211 Accession # P39905 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Astrocyte-derived trophic factor; Astrocyte-derived trophic factor ATF ATF1 ATF2 glial cell derived neurotrophic factor glial cell line derived neurotrophic factor glial cell line-derived neurotrophic factor HFB1-GDNF hGDNF HSCR3; ATF; Glial cell line-derived neurotrophic factor; hGDNF
Gene Aliases: ATF1; ATF2; GDNF; HFB1-GDNF; HSCR3
UniProt ID: (Human) P39905
Entrez Gene ID: (Human) 2668
Molecular Function: growth factor neurotrophic factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.